Anti-IRGM Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4015R-A488
10391-622EA 811.1 USD
10391-622
Anti-IRGM Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
IRGM belongs to a family of interferon-gamma inducible GTPases, belonging to the GTPase superfamily, and are selectively induced by IFN-gamma or bacterial lipopolysaccharide (LPS) stimulation. IRGM is primarily expressed in all cells derived from B-cell lineages, and is highly expressed in macrophages following IFN-gamma stimulation. IRGM has been shown to be required for host defences against a broad range of intracellular pathogens. Specifically, IRGM deficient mice show defects with intracellular microbial killing, phagosome maturation and autophagy. More recently IRGM has been shown to inhibit baseline hematopoietic proliferation and is required for a normal hematopoietic stem cell response to chemical and infectious stimuli.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IRGM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR