Anti-ATG9B Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4011R-CY7
10391-572EA 811.1 USD
10391-572
Anti-ATG9B Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Involved in autophagy and cytoplasm to vacuole transport (Cvt) vesicle formation. Plays a key role in the organization of the preautophagosomal structure/phagophore assembly site (PAS), the nucleating site for formation of the sequestering vesicle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATG9B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR