Anti-ATG9A Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-4010R-CY3
10395-550EA 811.1 USD
10395-550
Anti-ATG9A Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Involved in autophagy and cytoplasm to vacuole transport (Cvt) vesicle formation. Plays a key role in the organization of the preautophagosomal structure/phagophore assembly site (PAS), the nucleating site for formation of the sequestering vesicle. Cycles between a juxta-nuclear trans-Golgi network compartment and late endosomes. Nutrient starvation induces accumulation on autophagosomes. Starvation-dependent trafficking requires ULK1, ATG13 and SUPT20H.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATG9A
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR