Anti-ARSA Rabbit Polyclonal Antibody (Biotin)

BS-4006R-BIO
10391-484EA 529.29 USD
10391-484
Anti-ARSA Rabbit Polyclonal Antibody (Biotin)
Hydrolyzes cerebroside sulfate.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ARSA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR