Anti-ARSA Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-4006R-A647
10391-482EA 811.1 USD
10391-482
Anti-ARSA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Hydrolyzes cerebroside sulfate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARSA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR