Anti-SLC25A10 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3997R-A647
10391-314EA 811.1 USD
10391-314
Anti-SLC25A10 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Involved in translocation of malonate, malate and succinate in exchange for phosphate, sulfate, sulfite or thiosulfate across mitochondrial inner membrane.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC25A10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR