Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC16A7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3995R-A488
10394-404
Anti-SLC16A7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Tissues with few or no mitochondria, such as erythrocytes and tumor cells, depend largely on glycolysis to generate ATP. The major end products of glycolysis, pyruvate and lactate, must be eliminated from these cells to enable continued glycolytic flux and prevent toxic effects. H+/monocarboxylate transporters (MCTs) mediate the transport of lactate and pyruvate. Human MCT2 has a high affinity for the transport of pyruvate (summary by Lin et al., 1998 [PubMed 9786900]).[supplied by OMIM, Feb 2011].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC16A7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty