Anti-GYG1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3990R-CY5.5
10391-242EA 811.1 USD
10391-242
Anti-GYG1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
This gene encodes a member of the glycogenin family. Glycogenin is a glycosyltransferase that catalyzes the formation of a short glucose polymer from uridine diphosphate glucose in an autoglucosylation reaction. This reaction is followed by elongation and branching of the polymer, catalyzed by glycogen synthase and branching enzyme, to form glycogen. This gene is expressed in muscle and other tissues. Mutations in this gene result in glycogen storage disease XV. This gene has pseudogenes on chromosomes 1, 8 and 13 respectively. Alternatively spliced transcript variants encoding different isoforms have been identified.[provided by RefSeq, Sep 2010].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GYG1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR