Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GOT2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3985R-CY5.5
10395-466
Anti-GOT2 Rabbit Polyclonal Antibody (Cy5.5®)
GOT2 [Glutamate oxaloacetate transaminase] is a ubiquitous pyridoxal phosphate dependent enzyme which exists in both mitochondrial and cytosolic forms. The enzyme plays an important role in amino acid metabolism and in the urea and tricarboxylic acid cycles. The 2 isoenzymes are homodimeric. In liver about 80% of the enzyme activity is mitochondrial in origin, whereas in serum the enzyme activity is largely cytosolic. Although the mitochondrial and soluble forms of GOT are coded by different chromosomes, the 2 show close homology in amino acid sequence and were presumably derived from a common ancestral gene. Serum GOT [with SGPT] levels are usually elevated in states of hepatocellular injury (injury to the liver cells), the highest levels are associated with hepatitis of a viral origin. High levels are also found after myocardial infarction, when SGPT levels are lower.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GOT2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty