GOT2 [Glutamate oxaloacetate transaminase] is a ubiquitous pyridoxal phosphate dependent enzyme which exists in both mitochondrial and cytosolic forms. The enzyme plays an important role in amino acid metabolism and in the urea and tricarboxylic acid cycles. The 2 isoenzymes are homodimeric. In liver about 80% of the enzyme activity is mitochondrial in origin, whereas in serum the enzyme activity is largely cytosolic. Although the mitochondrial and soluble forms of GOT are coded by different chromosomes, the 2 show close homology in amino acid sequence and were presumably derived from a common ancestral gene. Serum GOT [with SGPT] levels are usually elevated in states of hepatocellular injury (injury to the liver cells), the highest levels are associated with hepatitis of a viral origin. High levels are also found after myocardial infarction, when SGPT levels are lower.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GOT2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat