Anti-FBP2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3981R-A647
10391-082EA 810.85 USD
10391-082
Anti-FBP2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
FBP2 is a gluconeogenesis regulatory enzyme which catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR