Anti-ENO3 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-3979R-CY7
10391-044EA 746.42 USD
10391-044
Anti-ENO3 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
ENO3 is also known as beta enolase, one of the three enolase isoenzymes found in mammals. A switch from alpha enolase to beta enolase occurs in muscle tissue during development and ENO3, a homodimer, is found in skeletal muscle cells in the adult and appears to have a function in striated muscle development and regeneration. Mutations can result in decreased stability of the enzyme and be associated with a glycogen storage myopathy. This results in exercise-induced myalgias, generalized muscle weakness and fatigability.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ENO3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR