Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ENO3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3979R-A555
10391-032EA 580.76 USD InStock
10391-032
Anti-ENO3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
ENO3 is also known as beta enolase, one of the three enolase isoenzymes found in mammals. A switch from alpha enolase to beta enolase occurs in muscle tissue during development and ENO3, a homodimer, is found in skeletal muscle cells in the adult and appears to have a function in striated muscle development and regeneration. Mutations can result in decreased stability of the enzyme and be associated with a glycogen storage myopathy. This results in exercise-induced myalgias, generalized muscle weakness and fatigability.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ENO3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty