Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AMY2A Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss

BS-3975R-BIO
10390-996EA 504.09 USD InStock
10390-996
Anti-AMY2A Rabbit Polyclonal Antibody (Biotin)
Antibodies
AMY2A belongs to the glycosyl hydrolase 13 family. Catalytic activity: endohydrolysis of alpha-D-glucosidic linkages in oligosaccharides and polysaccharides.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: AMY2A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty