Anti-SDHD Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3971R-CY7
10390-908EA 811.1 USD
10390-908
Anti-SDHD Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Membrane-anchoring subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDHD
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR