Anti-NDUFA8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3958R-A488
10390-670EA 811.1 USD
10390-670
Anti-NDUFA8 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NDUFA8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR