Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STXBP1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3954R-CY3
10390-598EA 580.76 USD InStock
10390-598
Anti-STXBP1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Munc18 may participate in the regulation of synaptic vesicle docking and fusion, possibly through interaction with GTP binding proteins. It is essential for neurotransmission and binds syntaxin, a component of the synaptic vesicle fusion machinery probably in a 1:1 ratio. Munc18 can interact with syntaxins 1, 2, and 3 but not syntaxin 4. It may play a role in determining the specificity of intracellular fusion reactions.Mutations in Munc18 can result in accumulations of acetylcholine and paralytic phenotype suggestive of a neurotransmitter release defect. Two Munc18 isoforms have recently been identified. Munc18-1 has been shown to be primarily expressed in brain, while Munc18-2 is expressed in most tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STXBP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty