Anti-STXBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3954R-A350
10390-588EA 811.1 USD
10390-588
Anti-STXBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Munc18 may participate in the regulation of synaptic vesicle docking and fusion, possibly through interaction with GTP binding proteins. It is essential for neurotransmission and binds syntaxin, a component of the synaptic vesicle fusion machinery probably in a 1:1 ratio. Munc18 can interact with syntaxins 1, 2, and 3 but not syntaxin 4. It may play a role in determining the specificity of intracellular fusion reactions.Mutations in Munc18 can result in accumulations of acetylcholine and paralytic phenotype suggestive of a neurotransmitter release defect. Two Munc18 isoforms have recently been identified. Munc18-1 has been shown to be primarily expressed in brain, while Munc18-2 is expressed in most tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STXBP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR