Anti-IDH2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3947R-CY5.5
10394-280EA 811.1 USD
10394-280
Anti-IDH2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Plays a role in intermediary metabolism and energy production. It may tightly associate or interact with the pyruvate dehydrogenase complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IDH2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR