Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GNG2 Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss

BS-3944R-BIO
10395-346
Anti-GNG2 Rabbit Polyclonal Antibody (Biotin)
GNG2 (Guanine nucleotide binding protein (G protein), gamma 2) is a member of the guanine nucleotide-binding proteins (G proteins) family. Heterotrimeric G proteins are involved as a modulator or transducer in various transmembrane signaling systems. The specificity of a G protein-receptor interaction is primarily mediated by the gamma subunit.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: GNG2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty