Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GNB3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-3943R-A488
10390-404
Anti-GNB3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
G proteins are composed of 3 units, alpha, beta and gamma and are involved in the modulation and transduction of various transmembrane signalling systems. There are families of related genes for each subunit. GNB3 encodes a beta subunit. Beta subunits are required for GTPase activity, for replacement of GDP by GTP, regulate alpha subunits and for G protein-effector interactions. A single-nucleotide polymorphism (C825T) in this gene is associated with essential hypertension and obesity and is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G protein beta 3/GNB3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty