Anti-GNB1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3941R-CY5
10390-366EA 810.85 USD
10390-366
Anti-GNB1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Heterotrimeric guanine nucleotide binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. GNB1 is a beta subunit. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G protein beta 1/GNB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR