Anti-GNAS Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3939R-CY3
10390-316EA 811.1 USD
10390-316
Anti-GNAS Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Guanine nucleotide-binding proteins (G proteins) are involved as modulators or transducers in various transmembrane signaling systems. The G(s) protein is involved in hormonal regulation of adenylate cyclase: it activates the cyclase in response to beta-adrenergic stimuli. Stimulates the Ras signaling pathway via RAPGEF2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G protein alpha S
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR