Anti-FH Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3938R-A647
10395-328EA 811.1 USD
10395-328
Anti-FH Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Defects in FH are the cause of fumarase deficiency (FHD)also known as fumaricaciduria. FHD is characterized by progressive encephalopathy, developmental delay, hypotonia, cerebral atrophy and lactic and pyruvic acidemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Fumarate hydratase/FH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR