Anti-COX3 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3934R-CY3
10390-234EA 811.1 USD
10390-234
Anti-COX3 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Cytochrome C Oxidase subunits I, II and III form the functional core of the enzyme complex.Defects in Cytochrome C Oxidase subunits III are a cause of Leber hereditary optic neuropathy (LHON) and cytochrome c oxidase deficiency (COX deficiency). Defects in Cytochrome C Oxidase subunits III are also found in mitochondrial encephalomyopathy with lactic acidosis and stroke-like episodes (MELAS) syndrome and recurrent myoglobinuria.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COX3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR