Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COX3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3934R-A647
10390-230
Anti-COX3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Cytochrome C Oxidase subunits I, II and III form the functional core of the enzyme complex.Defects in Cytochrome C Oxidase subunits III are a cause of Leber hereditary optic neuropathy (LHON) and cytochrome c oxidase deficiency (COX deficiency). Defects in Cytochrome C Oxidase subunits III are also found in mitochondrial encephalomyopathy with lactic acidosis and stroke-like episodes (MELAS) syndrome and recurrent myoglobinuria.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COX3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty