Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CACNA1E Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3933R-CY3
10390-208
Anti-CACNA1E Rabbit Polyclonal Antibody (Cy3®)
Calcium channels mediate the influx of calcium ions into the cell following membrane polarisation. R-type calcium channels such as Cav2.3 belong to the "high voltage-activated" group and are blocked by nickel. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Each of these proteins exists as multiple isoforms, either encoded by different genes or arising from alternative splicing of transcripts. Cav2.3 is an alpha-1 subunit and has 24 transmembrane segments, which form the pore through which ions pass into the cell. Calcium channels containing the Cav2.3 subunit may be involved in the modulation of firing patterns of neurons, which is important for information processing.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CACNA1E
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty