Calcium channels mediate the influx of calcium ions into the cell following membrane polarisation. R-type calcium channels such as Cav2.3 belong to the "high voltage-activated" group and are blocked by nickel. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Each of these proteins exists as multiple isoforms, either encoded by different genes or arising from alternative splicing of transcripts. Cav2.3 is an alpha-1 subunit and has 24 transmembrane segments, which form the pore through which ions pass into the cell. Calcium channels containing the Cav2.3 subunit may be involved in the modulation of firing patterns of neurons, which is important for information processing.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: CACNA1E
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat