Anti-CACNA1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-3930R-HRP
10395-960EA 811.1 USD
10395-960
Anti-CACNA1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Cav2.1 is a voltage-sensitive calcium channels (VSCC) which belongs to the calcium channel alpha-1 subunit family. Cav2.1 mediates the entry of calcium ions into excitable cells and is also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, gene expression, cell motility, cell division and cell death. Cav2.1 (isoform alpha-1A) gives rise to P and/or Q-type calcium currents. Voltage-dependent calcium channels are multisubunit complexes, consisting of alpha-1, alpha-2, beta and delta subunits in a 1:1:1:1 ratio. The channel activity is directed by the pore-forming and voltage-sensitive alpha-1 subunit. In many cases, this subunit is sufficient to generate voltage-sensitive calcium channel activity. The auxiliary subunits beta and alpha-2/delta linked by a disulfide bridge regulate the channel activity.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: CACNA1A
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR