Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CACNA1A Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss

BS-3930R-BIO
10390-182EA 504.09 USD InStock
10390-182
Anti-CACNA1A Rabbit Polyclonal Antibody (Biotin)
Antibodies
Cav2.1 is a voltage-sensitive calcium channels (VSCC) which belongs to the calcium channel alpha-1 subunit family. Cav2.1 mediates the entry of calcium ions into excitable cells and is also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, gene expression, cell motility, cell division and cell death. Cav2.1 (isoform alpha-1A) gives rise to P and/or Q-type calcium currents. Voltage-dependent calcium channels are multisubunit complexes, consisting of alpha-1, alpha-2, beta and delta subunits in a 1:1:1:1 ratio. The channel activity is directed by the pore-forming and voltage-sensitive alpha-1 subunit. In many cases, this subunit is sufficient to generate voltage-sensitive calcium channel activity. The auxiliary subunits beta and alpha-2/delta linked by a disulfide bridge regulate the channel activity.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: CACNA1A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty