Anti-ADCY8 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3925R-CY7
10390-110EA 811.1 USD
10390-110
Anti-ADCY8 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
This is a membrane-bound, calcium-stimulable adenylyl cyclase. May be involved in learning, in memory and in drug dependence (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADCY8
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR