Anti-ADCY5 Rabbit Polyclonal Antibody (Biotin)

BS-3922R-BIO
10390-052EA 529.29 USD
10390-052
Anti-ADCY5 Rabbit Polyclonal Antibody (Biotin)
This is a membrane-bound, calcium-inhibitable adenylyl cyclase.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ADCY5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR