Anti-STARD5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3919R-A555
10389-984EA 811.1 USD
10389-984
Anti-STARD5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
STARD5, or START domain containing 5, belongs to a family containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain. STARD5 may be involved in the intracellular transport of sterols or other lipids and may bind cholesterol or other sterols.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STARD5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR