Anti-ADCY10 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3916R-A647
10389-914EA 811.1 USD
10389-914
Anti-ADCY10 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Soluble adenylyl cyclase that has a critical role in mammalian spermatogenesis. Produces the cAMP which mediates in part the cAMP-responsive nuclear factors indispensable for maturation of sperm in the epididymis. Induces capacitation, the maturational process that sperm undergo prior to fertilization. May be the bicarbonate sensor. Involved in ciliary beat regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADCY10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR