Anti-SLCO2B1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3913R-CY3
10389-854EA 811.1 USD
10389-854
Anti-SLCO2B1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Mediates the Na(+)-independent transport of organic anions such as taurocholate, the prostaglandins PGD2, PGE1, PGE2, leukotriene C4, thromboxane B2 and iloprost.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLCO2B1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR