Anti-LGMN Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-3907R-CY5
10389-736EA 811.1 USD
10389-736
Anti-LGMN Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Has a strict specificity for hydrolysis of asparaginyl bonds. Can also cleave aspartyl bonds slowly, especially under acidic conditions. Required for normal lysosomal protein degradation in renal proximal tubules. Required for normal degradation of internalized EGFR. Plays a role in the regulation of cell proliferation via its role in EGFR degradation (By similarity). May be involved in the processing of proteins for MHC class II antigen presentation in the lysosomal/endosomal system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LGMN
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR