Anti-DIO3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-3902R-A555
10389-608EA 746.42 USD
10389-608
Anti-DIO3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Dio3 belongs to the iodothyronine deiodinase family. It has an essential role for regulation of thyroid hormone inactivation during embryological development in that it catalyzes the inactivation of thyroid hormone by inner ring deiodination of the prohormone thyroxine (T4) and the bioactive hormone 3,3',5-triiodothyronine (T3) to inactive metabolites, 3,3',5'-triiodothyronine (RT3) and 3,3'-diiodothyronine (T2), respectively. Dio3 may play a role in preventing premature exposure of developing fetal tissues to adult levels of thyroid hormones and has been linked to the development of consumptive hypothyroidism in both infants and adults.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIO3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR