Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DIO3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3902R-A350
10389-604EA 580.76 USD InStock
10389-604
Anti-DIO3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Dio3 belongs to the iodothyronine deiodinase family. It has an essential role for regulation of thyroid hormone inactivation during embryological development in that it catalyzes the inactivation of thyroid hormone by inner ring deiodination of the prohormone thyroxine (T4) and the bioactive hormone 3,3',5-triiodothyronine (T3) to inactive metabolites, 3,3',5'-triiodothyronine (RT3) and 3,3'-diiodothyronine (T2), respectively. Dio3 may play a role in preventing premature exposure of developing fetal tissues to adult levels of thyroid hormones and has been linked to the development of consumptive hypothyroidism in both infants and adults.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIO3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty