Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYP2R1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-3900R-A350
10389-580
Anti-CYP2R1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
CYP2R1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are mono-oxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a microsomal vitamin D hydroxylase that converts vitamin D into the active ligand for the vitamin D receptor.Defects in CYP2R1 are a cause of 25-hydroxyvitamin D(3) deficiency, also known as pseudovitamin D(3) deficiency rickets due to 25-hydroxylase deficiency. First described in patients who had rickets at a young age despite a history of adequate vitamin D intake. The patients sera had low calcium concentrations, low phosphate concentrations, elevated alkaline phosphatase activity and low levels of 25-hydroxyvitamin D.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP2R1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty