Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GTPBP10 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3893R-CY5.5
10389-426
Anti-GTPBP10 Rabbit Polyclonal Antibody (Cy5.5®)
May be involved in the ribosome maturation process. Complements an ObgE(CgtA) function in E.coli ribosome maturation. Plays a role of GTPase in vitro. When missing, disorganization of the nuceleolar architecture is observed.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GTPBP10
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty