Anti-GTPBP10 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3893R-A647
10389-418EA 811.1 USD
10389-418
Anti-GTPBP10 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
May be involved in the ribosome maturation process. Complements an ObgE(CgtA) function in E.coli ribosome maturation. Plays a role of GTPase in vitro. When missing, disorganization of the nuceleolar architecture is observed.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GTPBP10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR