Anti-NCF1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-3886R-CY5.5
10395-226EA 811.1 USD
10395-226
Anti-NCF1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
NCF2, NCF1, and a membrane bound cytochrome b558 are required for activation of the latent NADPH oxidase (necessary for superoxide production).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR