Anti-CYBA Rabbit Polyclonal Antibody

Supplier: Bioss

BS-3879R
10389-210EA 651.79 USD
10389-210
Anti-CYBA Rabbit Polyclonal Antibody
Antibodies
Critical component of the membrane-bound oxidase of phagocytes that generates superoxide. Associates with NOX3 to form a functional NADPH oxidase constitutively generating superoxide.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IHC-F(1:100-500; IF(IHC-P): 1:50-200; IF(IHC-F)(1:50-200

Type: Primary
Antigen: CYBA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR