Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ALOX12 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-3874R-HRP
10395-184
Anti-ALOX12 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
12 Lipoxygenase is a non heme iron containing dioxygenase which stereospecifically incorporates molecular oxygen into cis,cis-1,4-pentadiene containing polyunsaturated fatty acids. Murine leukocyte 12 Lipoxygenase oxidizes arachidonic acid to 12(S)-HpETE and 15(S)-HpETE in a ratio of 3:1. Murine leukocyte 12 Lipoxygenase is a 75 kDa protein which is 58% homologous to murine platelet 12 Lipoxygenase, and 70% homologous to porcine leukocyte 12 Lipoxygenase.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ALOX12
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty