Anti-ALOX12 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-3874R-CY5.5
10395-178EA 746.42 USD
10395-178
Anti-ALOX12 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
12 Lipoxygenase is a non heme iron containing dioxygenase which stereospecifically incorporates molecular oxygen into cis,cis-1,4-pentadiene containing polyunsaturated fatty acids. Murine leukocyte 12 Lipoxygenase oxidizes arachidonic acid to 12(S)-HpETE and 15(S)-HpETE in a ratio of 3:1. Murine leukocyte 12 Lipoxygenase is a 75 kDa protein which is 58% homologous to murine platelet 12 Lipoxygenase, and 70% homologous to porcine leukocyte 12 Lipoxygenase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALOX12
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR