Anti-GATA1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-3872R-CY7
10389-116EA 811.1 USD
10389-116
Anti-GATA1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GATA-1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR