Anti-PRR5 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-3866R-CY3
10389-006EA 811.1 USD
10389-006
Anti-PRR5 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Protor-1 plays an important role in regulation of PDGF receptor expression and in modulation of PDGF signaling. It is thought that Protor-1 may act as a tumor suppressor in breast cancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRR5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR