Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SCAP Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-3862R-A555
10388-952
Anti-SCAP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
SCAP is an escort protein required for cholestrol and lipid homeostasis. Cholesterol homeostasis in animal cells is achieved by regulated cleavage of SREBPs, membrane-bound transcription factors. SCAP forms a complex with SREBPs in order to release the active domains of SREBPs.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCAP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty