Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LPP Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3861R-A647
10388-930
Anti-LPP Rabbit Polyclonal Antibody (Alexa Fluor® 647)
LPP (LIM containing lipoma preferred partner), is a scaffolding protein which contains three LIM domains at its carboxy terminus, preceded by a proline rich pre LIM region containing a number of protein interaction domains. LPP localizes to sites of cell adhesion, such as focal adhesions and cell-cell contacts and may be involved in cell-cell adhesion and cell motility. LPP also shuttles through the nucleus and may function as a transcriptional co-activator. The human LPP gene maps to chromosomal location 3q28, and preferentially translocates to the HMGIC gene in a subclass of human benign mesenchymal tumors known as lipomas. Alternate splicing results in multiple transcript variants.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LPP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty