Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CHRDL2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-3860R-A647
10388-906
Anti-CHRDL2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
CHRDL2 is a novel chordin like protein that can act as a BMP antagonist. Amember of the chordin family of proteins, it contains a signal peptide andthree CR (cysteine-rich repeat) domains. When expressed as a recombinantprotein it is secreted and binds to activin A, but not to BMP-2, -4, -6. Differential expression has been detected in developing chondrocytes, myoblasts, osteoblasts, and osteoarthritic joints. Complex alternative splicing of CHRDL2 potentially results in distinct isoforms that differ at their C termini, in the expression of signal peptide, and in the content of CR domains. CHRDL2 was originally characterized as a novel protein exclusively expressed in breast, lung, and colon tumors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHRDL2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty